Learn More
Macherey-Nagel™ Alugram Aluminum Sheet Nano-SIL G
Brand:  Macherey-Nagel™ 818241
 View more versions of this product
Product Code. 12774738
  • € 278.00 / Pack of 25
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
2 to 10 μm
25 Pk.
0.75 mL/g
0.20 mm
60 Å
Aluminum Sheet
∼500 m2/g
20 x 20 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Alugram Aluminum Sheet Nano-SIL G > Plate Size: 200 x 200mm