missing translation for 'onlineSavingsMsg'
Learn More

Aalborg™ Aluminum Flowmeter Frame for 150mm P-Meter

Aalborg™ Aluminum Flowmeter Frame for 150mm P-meter is for use with single tube flow meters, which are available with built in valves.

Brand:  Aalborg™ P11A2-BA0

Product Code. 11787699

  • € 127.00 / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Rib-guided or fluted metering tubes facilitate stable and accurate readings
  • Magnifier lens in front shield to enhance reading resolution
  • Interchangeability of flowtubes and floats
  • Ease of installation and exchange of flow tubes
  • Glass flow tubes are prevented from turning during the tightening phase of the assembly procedure
  • Chemical compatibility
  • Simple means of panel mounting
  • Frame for P-meter
Specifications

Specifications

Single Tube P-Meter
Alu Flowmeter
150 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.