Learn More
Ansell AlphaTec™ 38-628 Butyl/Viton Immersion Gloves
AlphaTec™ Gloves 38-628 offer a combination of butyl and viton layer of 0.7 mm for a maximum protection against hazardous chemicals. Provide the best resistance to the most aggressive chemicals without compromising dexterity or comfort. Offer superior protection against aliphatic, halogenated and aromatic hydrocarbons (benzene, toluene, xylene), as well as concentrated mineral acids.
Brand:  Ansell 38-628_10
 View more versions of this product
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15252319
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Beschrijving
Beschrijving
  • Surprisingly flexible and provides excellent dexterity
  • Designed for comfort with a natural, curved ergonomic shape and feel
  • Offers easy donning and a good grip
  • Absence of latex means there's no risk of latex allergies
  • Ideal for the chemical, automotive and printing industries, oil and petrol refining, mining, aerospace and disaster response
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
Viton/Butyl
Viton/Butyl Polymer
Yes
10
28 mil
Beaded/Rolled
Yes
1 Pair
38-628
Viton/Butyl
Yes
Immersion Gloves
38-628
Black
Black
Individually wrapped bags with insert
Highly chemical resistance
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Speciale aanbiedingen
Speciale aanbiedingen
Product Content Correction
Product Title
Ansell AlphaTec™ 38-628 Butyl/Viton Immersion Gloves > Size 10