missing translation for 'onlineSavingsMsg'
Learn More

Allyl N-[2-(methylamino)ethyl]carbamate , Tech., Thermo Scientific™

Product Code. 10225794
Click to view available options
Amber Glass Bottle; 1g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 188403-10-3
Molecular Formula C7H14N2O2
MDL Number MFCD08690326
Packaging Amber Glass Bottle
Synonym allyl n-2-methylamino ethyl carbamate, prop-2-en-1-yl 2-methylamino ethyl carbamate, prop-2-en-1-yl n-2-methylamino ethyl carbamate, allyl 2-methylamino ethyl carbamate, n-2-methylamino ethyl prop-2-enyloxycarboxamide, carbamic acid, 2-methylamino ethyl-, 2-propenyl ester 9ci
InChI Key KWUXKYITOPQQRK-UHFFFAOYSA-N
SMILES CNCCNC(=O)OCC=C
IUPAC Name prop-2-enyl N-[2-(methylamino)ethyl]carbamate
Molecular Weight (g/mol) 158.201
PubChem CID 18525944
Formula Weight 158.2
Grade Technical
Quantity 1 g
Chemical Name or Material Allyl N-[2-(methylamino)ethyl]carbamate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'