missing translation for 'onlineSavingsMsg'
Learn More
Alloxan monohydrate, 98%, Thermo Scientific Chemicals Product Code.: 10511661

Alloxan monohydrate, 98%, Thermo Scientific Chemicals

Product Code. 10511661
100 g, Glass bottle
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10511661

Brand: Thermo Scientific Acros 215371000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3237-50-1, 50-71-5
Molecular Formula C4H4N2O5
MDL Number MFCD00149399
InChI Key ZIIHZVKHFWOENY-UHFFFAOYSA-N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Beige-Yellow to White
Assay Percent Range 98%
Beilstein 24, 500
Merck Index 10, 276
Synonym isovaleraldehyde, 2,4-dinitrophenyl hydrazone, 5,5-dihydroxyperhydropyrimidinetrione, 5,5-dihydroxybarbituric acid, 5,5-dihydroxy-2,4,6 1h,3h,5h-pyrimidinetrione, ccris 5957, butanal, 3-methyl-, 2,4-dinitrophenyl hydrazone, isovaleraldehyde 2,4-dinitrophenyl hydrazone, n-e-3-methylbutylideneamino-2,4-dinitroaniline, isovaleraldehyde, 2,4-dinitrophenylhydazone, butyraldehyde, 3-methyl-, 2,4-dinitrophenyl hydrazone, isovaleraldehyde dnp, o2aap9f8b6, unii-o2aap9f8b6, mesoxalylurea monohydrate, 2,4,5,6 1h,3h-pyrimidinetetrone hydrate, 2,4,6 1h,3h,5h-pyrimidinetrione, 5,5-dihydroxy, barbituric acid, 5,5-dihydroxy, 1e-3-methylbutanal 2,4-dinitrophenyl hydrazone #, isovaleraldehyd-2,4-dinitrophenylhydrazone, isovaleraldehyde,4-dinitrophenyl hydrazone
SMILES OC1(O)C(=O)NC(=O)NC1=O
IUPAC Name 5,5-dihydroxy-1,3-diazinane-2,4,6-trione
Molecular Weight (g/mol) 160.09
PubChem CID 9569524
Formula Weight 160.09
Percent Purity ≥97.5%
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Powder
Chemical Name or Material Alloxan monohydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Alloxan monohydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.