Learn More
Allihn Condenser with Ground Cone and Socket
Product Code. p-8319008
Click to view available options
Standard Taper Joint No:
24/29
29/32
Length (Metric) Jacket:
545 mm
550 mm
Unit Size:
Each
Pack of 2
Product Code. 11381845
Brand: Quickfit CX7/33/SC
Only null left
Add to basket
Add to basket
Ideal for reflux applications.
  • Inner bulb sections provide increased cooling surface
  • With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Ideal for reflux work
Length (Metric) Jacket 545 mm
Length (Metric) 545 mm
Standard Taper Joint No 24/29
Material Borosilicate Glass
Type Allihn Condenser