Learn More
Al-23 Solid Rod, Round, Dia.: 4mm, Thermo Scientific™
€ 22.00 - € 121.00
Specifications
Chemical Name or Material Al-23 Solid Rod
Form Rod
View More Specs
Products 7
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11363068
View Documents
Thermo Scientific Alfa Aesar
032929.DM
100 mm N/A N/A N/A
11393068
View Documents
Thermo Scientific Alfa Aesar
032929.DR
200 mm N/A N/A N/A
11323078
View Documents
Thermo Scientific Alfa Aesar
032929.DT
250 mm N/A N/A N/A
11303078
View Documents
Thermo Scientific Alfa Aesar
032929.DW
300 mm N/A N/A N/A
11373068
View Documents
Thermo Scientific Alfa Aesar
032929.EH
600 mm N/A N/A N/A
11353068
View Documents
Thermo Scientific Alfa Aesar
032929.EM
800 mm N/A N/A N/A
11313078
View Documents
Thermo Scientific Alfa Aesar
032929.EO
1000 mm N/A N/A N/A
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Al-23 Solid Rod
Rod
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Al-23 Solid Rod, Round, Dia.: 4mm, Thermo Scientific™