Learn More
Aalborg™ Air Mass Flowmeter
Brand:  Aalborg™ GFM17A-VAL6-A0/3AB-08-SS
 View more versions of this product
Product Code. 11832220
  • € 1040.00 / Each
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Specifications
Mass Flowmeter
0 to 2 LPM
12 V to 200 mA max
Full scale
±5%
NIST calibration certificate
±1.5 %
5°C to 50°C