Learn More
Abeba™ Air-Cushion 32310 Shoes
Air cushion PU-sole with high slip resistance. Sanitized™ treated.
Brand:  Abeba™ 32310/46
71.25 EUR valid until 2025-02-13
Use promo code "24130" to get your promotional price.
 View more versions of this product
Product Code. 15659657
  • € 96.70 / 1 pair
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20347:2012 O1 SRA
  • Smooth leather, perforated
  • Black
  • Slip resistance PU outsole sole
  • Replaceable insole (article no. 3551)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Instep area with elastic band
Specifications
Specifications
CE EN ISO 20347:2012 O1 SRA
Microfiber
46
Polyurethane
Black
Leather
Slip-resistant
32310
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers