Learn More
AIM2 Mouse anti-Human, Unconjugated, Clone: 3E12A8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66902-1-IG-150UL
 View more versions of this product
Product Code. 16819185
  • € 422.30 / 150µL
Description
The AIM2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AIM2. This antibody recognizes human, mouse, rat, rabbit antigen. The AIM2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
AIM2
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Rabbit
Western Blot
3E12A8
Unconjugated
150 μL
9447