Learn More
AIM-100, MedChemExpress
Product Code. 16446716
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16446716
Brand: Medchem Express HY15290
MedChemExpress AIM-100 is a potent and selective Ack1 inhibitor with an IC50 of 21.58 nM. AIM-100 also inhibits Tyr267 phosphorylation. AIM-100 does not inhibits other kinases including PI3-kinase and AKT subfamily members. AIM-100 has an anticancer effect.
AIM-100 is a potent and selective Ack1 inhibitor with an IC50 of 21.58 nM. AIM-100 also inhibits Tyr267 phosphorylation. AIM-100 does not inhibits other kinases including PI3-kinase and AKT subfamily members. AIM-100 has an anticancer effect.
Specifications
Chemical Name or Material AIM-100
CAS 873305-35-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H21N3O2
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (134.61 mM; Need ultrasonic)
SMILES C1(C2=C(C3=CC=CC=C3)OC4=C2C(NC[C@@H]5CCCO5)=NC=N4)=CC=CC=C1
Molecular Weight (g/mol) 371.43
Formula Weight 371.43
Percent Purity 99.92%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only