missing translation for 'onlineSavingsMsg'
Learn More

Scharlab Agar-Agar, Powder, For Bacteriology

Product Code. 12407941
This item is not returnable. View return policy

Product Code. 12407941

Brand: Scharlab AG00200500

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Used in bacterial culturing. Scharlab Agar-Agar Powder, For Bacteriology is a necessary ingredient for the preparation of nutrient media. It is also used in the manufacture of dyes and as an emulsifier.

  • CAS: 9002-18-0
  • EIN ECS No.: 232-658-1
  • pH in gel (1.5%, after autoclaving): 6.0–7.5
  • Gel point (1.5 %, after autoclaving): 34–36°C
  • Gel strength (1.5%, after autoclaving): 650–750 Nikan
  • Nephelometry (after autoclaving): max. 10 N.T.U
  • Phosphates (as PO4): max. 0.1%
  • Calcium (Ca) : max. 0.1%
  • Magnesium (Mg): max. 0.05%
  • Full ash: max. 6%
  • Loss on drying: max. 8%

Specifications

Form Powder
Quantity 500 g
Synonym Agar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.