Learn More
B Braun™ Aesculap™ Safety Scalpel
Product Code. 15303515
Click to view available options
Size:
# 11
# 13
# 15
# 20
# 36
Unit Size:
Pack of 10
Product Code. 15303515
Brand: B Braun™ BA813SU
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Reduces the risk of infections caused by cuts and stab wounds suffered during handover, take-over and disposal of scalpels.
  • Safety locking mechanism for disposal
  • Single-handed operation
  • Ergonomic guide for precise incision
  • Ruler in cm and inches
  • Transparent cap for blade protection – blade and position is always visible
  • Provides optimal sharpness
Specifications
Blade Material Carbon Steel
Size # 13
Description Surgeon Grade
Disposable Single-use
Sterility Sterile
Includes Protective Cap
Handle Material Plastic
Quantity 10/Pk.