Learn More
Eppendorf™ Adaptor
Shop All Eppendorf Products
Click to view available options
Thread Size:
38 mm
40 mm
Unit Size:
Each
Specifications
Specifications
| Product Type | Adapter |
| For Use With (Equipment) | Bottle |
| Quantity | 1 Ea. |
| Thread Size | 40 mm |