missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Adapters, set of 2
Designed for use with Eppendorf rotors
Brand: Eppendorf™ 5920711008
319.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
for 50mL, set of 2 pcs.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
for Eppendorf Tubes™ 5.0mL and 15mL | |
5.0 and 15 mL Eppendorf Tubes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction