Learn More
Fisherbrand™ Adapters for Fisherbrand™ accuSpin™ 4-Place Rotor with Rectangular Bucket
Product Code. 10371092 Shop All Thermo Scientific Products
Click to view available options
Container Type:
Bottle
Color:
Brown
Gray
White
Unit Size:
Each
Product Code. 10371092
Brand: Fisherbrand™ 75005330
Adapters are sold individually and are available in other sizes for convience.
Fit in rectangular bucket 13-100-587 for swinging bucket rotor 13-100-584; also can be used in sealed rotor 13-100-589.
Order Info
Adapters are sold individually. Order correct quantity to outfit rotor. Other adapter sizes are available online.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) accuSpin 1/1R 4-Place Rotor
Color Gray
Container Type Bottle