Learn More
Eppendorf™ Adapter for S-4x400 Bucket
Shop All Eppendorf Products
Click to view available options
Capacity:
1 x 175 to 225 mL Conical Tubes
1 x 175 to 250 mL Flat/Conical Bottle
1 x 400 mL Bottle
11 x 7.5 to 12 mL Round Bottom Tubes
15 x 4 to 8 mL Round Bottom Tubes
17 x 12 x 75 mm Round Bottom Tubes
26 x 1.5/2.0 mL Microtubes
4 x 50 mL Conical Tubes
50 mL Self-standing Tubes, 50 mL Centriprep, 30 mL Oak Ridge
7 x 5.0 mL Microtubes, 15 mL Conical Tubes
8 x 9 mL Round Bottom Tubes
Unit Size:
Pack of 2
Description
- Suitable for S-4x400 Rotor buckets
- Accommodates selected array of tubes or bottles
Specifications
Specifications
| Tube Type | Flat/Conical Bottom |
| Capacity | 1 x 175 to 250 mL Flat/Conical Bottle |
| Max. Tube Diameter | 62 mm |
| Max. Tube Length | 138 mm |
| For Use With (Equipment) | S-4x400 Bucket |
| Vessels per Adapter | 1 |
| No. of Places | 1 |
| No. per Pack | 2/Pk. |