Learn More
Eppendorf™ Adapter for S-4x400 Bucket
Product Code. 15813222 Shop All Eppendorf Products
Click to view available options
Capacity:
1 x 175 to 225 mL Conical Tubes
1 x 175 to 250 mL Flat/Conical Bottle
1 x 400 mL Bottle
11 x 7.5 to 12 mL Round Bottom Tubes
15 x 4 to 8 mL Round Bottom Tubes
17 x 12 x 75 mm Round Bottom Tubes
26 x 1.5/2.0 mL Microtubes
4 x 50 mL Conical Tubes
50 mL Self-standing Tubes, 50 mL Centriprep, 30 mL Oak Ridge
7 x 5.0 mL Microtubes, 15 mL Conical Tubes
8 x 9 mL Round Bottom Tubes
Unit Size:
Pack of 2
Product Code. 15813222
Brand: Eppendorf™ 5910701004
Facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for S-4x400 rotor.
  • Suitable for S-4x400 Rotor buckets
  • Accommodates selected array of tubes or bottles
TRUSTED_SUSTAINABILITY
Specifications
Tube Type Conical
Capacity 4 x 50 mL Conical Tubes
Max. Tube Diameter 29 mm
Max. Tube Length 125 mm
For Use With (Equipment) S-4x400 Bucket
Vessels per Adapter 4
No. of Places 4
No. per Pack 2/Pk.