Learn More
Socorex™ Acura™ Manual 815 Fixed Volume Micropipettes
Product Code. 10733463
Click to view available options
Volume (Metric):
1 μL
5 μL
10 μL
15 μL
20 μL
25 μL
40 μL
50 μL
60 μL
70 μL
75 μL
80 μL
90 μL
100 μL
120 μL
150 μL
200 μL
250 μL
300 μL
400 μL
500 μL
600 μL
700 μL
750 μL
800 μL
900 μL
1000 μL
Unit Size:
Each
Product Code. 10733463
Brand: Socorex™ 815.1000
Only null left
Add to basket
Add to basket
Slim and precise micropipettes set at a fixed volume for easy dose repetition. Socorex™ Acura™ Manual 815 Fixed Volume Micropipettes are easy to handle and have a soft plunger stroke.
  • Air-displacement fixed-volume pipette.
    • Exceptional, user-friendly ergonomics.
    • Justip™ adjustable tip ejector fitting most tips.
    • Shock, UV-light and autoclaving resistance.
    • Ultra-soft plunger stroke.
    • Available in a variety of volume models.
Specifications
Accuracy <±0.6%
Certifications/Compliance ISO 8655, GMP, GLP, NCCLS, CE IVD Directive 98/79 EEC (in vitro diagnostic medical devices).
Autoclavable Autoclavable
Color Blue
Model 815
Type Micropipette
Volume (Metric) 1000 μL
Precision <0.2%
Quantity 1 Each
Warranty 36 Months