missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Socorex™ Acura™ Electro 956.08 Micropipette Initial Kit
Ergonomic electronic multi-channel micropipette initial package. Socorex™ Acura™ Electro 956.08 Micropipette Initial Kit comes with pipette, pipette charging stand, fast charger with AC adapter and cord, additional battery pack, and pipette tip samples.
Brand: Socorex™ 956.08.010E
This item is not returnable.
View return policy
Description
- Exceptional, user-friendly ergonomics.
- Lightweight.
- Easy, intuitive, self-teaching program.
- Large display, reversible right/left reading.
- Microprocessor-controlled motor.
- Swift-interchange, rapidly charging, long-working, rechargeable battery pack.
- Sequential tip ejection.
- Also fit any single channel volumetric module.
Specifications
<±3.5% - <±1.0% | |
EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS) | |
Acura™ 956 | |
Acura™ | |
0.5 to 10 μL | |
Pipette charging stand, fast charger with AC adapter and cord, additional battery pack, QC certificate, Qualitips™ pipette tip samples and operating instructions. | |
<3.0% - <0.7% | |
<1.5Hours |
4.8V NiMH | |
Single-Channel Volumetric Module | |
8 | |
Electronic Micropipette | |
Right/Left LCD | |
0.05μL | |
1 Each | |
24 Months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction