Learn More
Cytiva AcroPak™ 800 Capsules with Fluorodyne™ II Membrane
Product Code. 17154241
Click to view available options
Pore Size:
0.1 μm
0.2 μm
Unit Size:
Each
Product Code. 17154241
Brand: Cytiva 12473
AcroPak 800 Capsules with Fluorodyne II (PVDF) membrane that is compatible with aqueous and many organic solvents.
  • Double layer sterilizing membrane enhances reliability
  • Upstream air vent prevents vapor lock
  • Manufactured without the use of adhesives to minimize extractables
  • Pre-sterilized by gamma irradiation
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) Hydrophilic PVDF
Pore Size 0.1 μm
Sterility Sterile
Filtration Area 800 cm2
Outlet Style 9/16 in. Hose Barb
Quantity 1/Ea.