Learn More
ACP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11594-1-AP-20UL
 View more versions of this product
Product Code. 16878773
  • € 145.00 / 20µL
Description
Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher′s disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin′s lymphoma and in the sera of patients undergoing active bone turnover.
Specifications
Specifications
ACP5
Polyclonal
Unconjugated
ACP5
ACP5, Tartrate resistant acid ATPase, TR AP, TRACP, TRAP, TrATPase, Type 5 acid phosphatase
Rabbit
Antigen Affinity Chromatography
RUO
11433, 25732, 54
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P13686, P29288, Q05117
ACP5
ACP5 Fusion Protein Ag2065
20 μL
Primary
Mouse, Human, Rat