missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, Puriss., meets analytical spec. of BP, NF, Ph. Eur., ≥99% (GC), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
20 kg
25 L
155 kg
Packaging:
Glass Bottle
Metal Drum
Plastic Bottle
Stainless Steel Can
Unit Size:
1 L
10 L
155kg
2.5 L
20kg
25 L
5 L
Specifications
Specifications
| Density | 0.791g/mL at 25°C, 0.790 to 0.792g/mL at 20°C |
| Quantity | 10 L |
| Explosion Limit | 0.132 |
| Flash Point | −17°C (1.4°F) |
| Packaging | Stainless Steel Can |
| Linear Formula | CH3COCH3 |
| Vapor Pressure | 184mmHg (20°C) |
| UN Number | 1090 |
| Beilstein | 635680 |
| Formula Weight | 58.08g/mol |
| Show More |
Safety and Handling
Product Identifier
- Acetone
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction