missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid Solution 50%, Chem Lab™

Product Code. 11979152
Click to view available options
Quantity:
5 L
Unit Size:
5 L
This item is not returnable. View return policy

Product Code. 11979152

Brand: Chem-Lab CL02.0133.5000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

pH <2
Boiling Point 100°C
Color Colorless
Solubility Information Soluble in water
Density 1.03g/mL
Quantity 5 L
Odor Specific
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Acetic acid 50 vol. % solution
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Acetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.