missing translation for 'onlineSavingsMsg'
Learn More

Acetazolamide, 99%

Product Code. 11381947
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11381947

Brand: Thermo Scientific Alfa Aesar L07562.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59-66-5 | C4H6N4O3S2 | 222.24 g/mol

Acetazolamide acts as a carbonic anhydrase inhibitor which increases cerebral blood flow. It inhibits water permeability of membranes by interacting with aquaporins. It is used for the medical treatment of glaucoma, epileptic seizure, idiopathic intracranial hypertension, altitude sickness, cystinuria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in NH4OH (50 mg/mL), DMSO, Methanol and slightly soluble in Ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-66-5
Molecular Formula C4H6N4O3S2
Molecular Weight (g/mol) 222.24
MDL Number MFCD00003105
InChI Key BZKPWHYZMXOIDC-UHFFFAOYSA-N
Synonym acetazolamide, diamox, acetazolamid, diacarb, glaupax, defiltran, nephramide, acetamox, cidamex, diluran
PubChem CID 1986
ChEBI CHEBI:27690
IUPAC Name N-(5-sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide
SMILES CC(=O)NC1=NN=C(S1)S(N)(=O)=O

Specifications

Melting Point ∽255°C (decomposition)
Quantity 25 g
Beilstein 212994
Merck Index 14,53
Solubility Information Soluble in NH4OH (50mg/mL),DMSO,Methanol and slightly soluble in Ethanol.
Formula Weight 222.25
Percent Purity 99%
Chemical Name or Material Acetazolamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.