Learn More
Thermo Scientific™ Accessories for RT Basic Series and RT Touch Series Magnetic Stirrers
Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric):
170 mm
220 mm
Type:
Plate Cover
Unit Size:
Each
Description
Use these Thermo Scientific™ Accessories for RT Basic Series Magnetic Stirrers to optimize performance of your stirrer.
Specifications
Specifications
| Color | White |
| Diameter (Metric) | 220 mm |
| For Use With (Equipment) | RT Basic Series and RT Touch Series Magnetic Stirrers |
| Holds | 96-Well Plates, 384-Well Plates |
| Type | Plate Cover |
| Material | Silicone |