Learn More
Thermo Scientific™ Accessories for MaxQ™ HP Orbital Shakers
Shop All Thermo Scientific Products
Click to view available options
Product Type:
7 Day Recorder, 220V
Gassing Manifold (One Gas)
Magnet and Pen Kit
Platform Adapter
Shaker Kit with Mount and Stand
For Use With (Equipment):
MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000
MaxQ HP 416, 430, 420
MaxQ HP 416, 430, 435, 436, 480, 481
MaxQ HP 420
MaxQ HP 420, 435, 436, 480, 481
MaxQ HP 435, 436, 480, 481
Unit Size:
Each
Description
Get maximum flexibility, regardless of your application, by using Thermo Scientific™ Accessories for MaxQ™ High Performance Orbital Shakers.
Specifications
Specifications
| Electrical Requirements | 220 V, 50/60 Hz |
| Voltage | 220 V |
| Temperature Range (Metric) | 0°C to 100°C |
| Product Type | 7 Day Recorder, 220V |
| Frequency | 50/60 Hz |
| For Use With (Equipment) | MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000 |