Learn More
Fisherbrand™ Accessories for Fisherbrand™ Multi Platform Shaker
Accessories required for the use of Fisher Scientific™ Multi Platform Shaker
Brand:  Fisherbrand™ 88861132
179.65 EUR valid until 2025-01-31
Use promo code "24160" to get your promotional price.
 View more versions of this product
Product Code. 15534100
  • € 193.00 / Each
Estimated Shipment: 20-08-2025
to see stock.
Explore more special offers
Description
Accessories for multi platform shaker to enhance performance and flexibility
  • Increase performance and flexibility
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Platform with Clamps for 6 x 250mL flasks/150mL beakers