Learn More
BINDER™ Access Port with Silicone Plug for Series CB, CBF
BINDER™ Access Port with silicone plug is an option for use with CO2 incubators CB and CBF
Brand: BINDER™ 8612-0026
Description
- Access Port
- With Silicone Plug
- For use with Constant Climate Chambers KBF-S and KBF-S ECO
Specifications
Access port with silicone plug - 30 mm, left | |
Silicone | |
Left Position | |
1.2 in. |
BINDER Series CB and CBF | |
Access Port | |
Silicone plug | |
30 mm |