missing translation for 'onlineSavingsMsg'
Learn More

AC 261066, Tocris Bioscience™

Product Code. 18024042
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

RARβ2 agonist

Potent RARβ2 agonist (pEC50 = 8.1). Selective over RARβ1, RARα and RARγ (pEC50 values are 6.4, 6.2 and 6.3 respectively). More potent and orally available than AC 55649 (Cat. No. 2436).

Chemical Identifiers

CAS 870773-76-5
Molecular Formula C17H20FNO4S
Molecular Weight (g/mol) 353.408
InChI Key HSAOETBFVAWNRP-UHFFFAOYSA-N
Synonym unii-lgf3f89bzp, lgf3f89bzp, 4-4-2-butoxyethoxy--5-methyl-2-thiazolyl-2-fluorobenzoic acid, benzoic acid, 4-4-2-butoxyethoxy-5-methyl-2-thiazolyl-2-fluoro, 4-4-2-butoxyethoxy-5-methyl-1,3-thiazol-2-yl-2-fluorobenzoic acid, d09rrf, 4-4-2-butoxyethoxy-5-methylthiazol-2-yl-2-fluorobenzoic acid
PubChem CID 11530459
IUPAC Name 4-[4-(2-butoxyethoxy)-5-methyl-1,3-thiazol-2-yl]-2-fluorobenzoic acid
SMILES CCCCOCCOC1=C(SC(=N1)C2=CC(=C(C=C2)C(=O)O)F)C

Specifications

Quantity 10 mg
Formula Weight 353.41
Percent Purity >98%
Chemical Name or Material AC 261066
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'