Learn More
Uvex™ 9790 Chinstraps
Product Code. p-7221941 Shop All Uvex Products
Click to view available options
Material:
Elastic
Unit Size:
Each
Product Code. 17289191
Brand: Uvex™ 9790031
Only null left
Add to basket
Add to basket
Chinstraps for use with Uvex™ Safety Helmets available in various materials.
  • For use with Uvex™ Safety Helmets
TRUSTED_SUSTAINABILITY
Specifications
Color Gray, Green
For Use With (Equipment) Pheos Safety Helmets
Material Elastic
Model 9790
Product Type Chin Strap