Learn More
uvex 9169 Super F OTG CR Safety Spectacles
Shop All Uvex Products
Click to view available options
:
Clear
Description
- Designed to provide professinal protection in laboratories, clinics, cleanrooms, or in food production
- Spectacles are capable of withstanding multiple autoclave sterilization cycles
- Can be worn over perscription lenses, making them perfect for visitors
- Certified to EN 166 (personal eye protection) and EN 170 (UV filters)
- Uvex™ Supravision Clean lens coating features a chemical-resistant, anti-fog interior and an extremely scratch-resistant exterior
- Soft arm ends provide additional comfort
- Unrestricted peripheral vision
Specifications
Specifications
| Lens Type | Supravision Clean |
| Lens Coating | Anti-fog |
| Certifications/Compliance (Europe) | EN 166:2001, EN 170:2002 |
| Lens Tint | Clear |
| Product Type | Overglasses |
| Frame Color | White, Blue |
| Material | Polycarbonate |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Frame Material | Plastic |
| Show More |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion