Learn More
Fisherbrand™ 8mm Push On Cap, for 8mm Crimp Vials, thinned penetration area
Polyethylene caps are chemically inert and suitable for most chromatography applications.
Brand: Fisherbrand™ 08 08 1675
This item is not returnable.
View return policy
Description
- 8mm PE Push-On Cap
- Blue
- Thinned penetration area
- Snap caps are easy to apply and easy to remove, eliminate the need for crimping or de-capping tools
Specifications
Blue | |
Polyethylene | |
Push On Cap |
8 mm | |
100 | |
ND8 |