Learn More
Tuttnauer 85L Vertical Autoclave
The 3870ELV-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.
Brand: Tuttnauer 3870ELVCPVGP-380-D
Description
Features
- Fo software control
- PID (Proportional Integral Differential) pressure control
- Stores the last 200 cycles in built-in memory
- Two independent flexible PT100 temperature sensors to prevent over-boiling of liquids and explosions of bottles
- Up to 6 temperature sensors and 4 pressure sensors can be connected
- 30 Identification Codes and Passwords for access level control
- The controller and software comply with the 21 CFR part 11 standard
Innovative Multi-Color Display
- Multi-color display for easier reading
- Color is used to indicate the stage of the cycle
- Quick access to important information
- 26 Languages
- Built-in view of historical cycle data
Specifications
18.7 gal. | |
70 cm | |
28 in. | |
28 in. | |
Autoclave, Top-loading | |
105°C to 138°C | |
316L Stainless Steel |
85 L | |
15.2 in. | |
38 cm | |
70 cm | |
221°F to 280°F | |
Three Phase, Cooling, Vacuum, Steam Generator, Printer | |
Non-sterile |