Learn More
Brady™ 7960 Series Thermal Transfer Printer Ribbon
Thermal transfer ribbon with excellent smearing resistance and good solvent resistance.
Brand:  Brady™ 236010
 View more versions of this product
Product Code. 11508724
  • € 306.00 / Each
In Stock
to see stock.
Explore more special offers
Description
When printed on the appropriate Brady™ label, this ribbon gives good solvent resistance.
  • Lower for alcohol
  • High print quality
  • Excellent smear resistance
Specifications
Specifications
Black
Resin
6.7 in.
170 mm
25.4 mm
2011/65/EU and 2015/863
BradyPrinter PR Plus Series Label Printer
984 ft.
300 m
1 in.
Open-core Ribbon
R7960-170X300/O