Learn More
Brady™ 6000 Series Halogen-free Thermal Transfer Printer Ribbon
Thermal transfer ribbon with excellent solvent, high heat, and super scratch resistance.
Brand: Brady™ 196509
This item is not returnable.
View return policy
Description
High density halogen-free ribbon, excellent solvent resistance, high heat resistance, and super scratch resistance.
Specifications
Black | |
Resin | |
3.3 in. | |
83 mm | |
25.4 mm |
BradyPrinter i7100 Industrial Label Printer, BradyPrinter PR Plus Series Label Printer | |
984 in. | |
300 m | |
1 in. | |
2011/65/EU and 2015/863 |