missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™

Product Code. 15429045
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 749849-14-7
Molecular Formula C8H5BrN2O2
Molecular Weight (g/mol) 241.04
MDL Number MFCD04116918
InChI Key FQBFPVLZLNEQOE-UHFFFAOYSA-N
Synonym 6-bromoimidazo 1,2-a pyridine-2-carboxylic acid, 6-bromo-imidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylic acid, 6-bromo, 6-bromo-4-hydroimidazo 1,2-a pyridine-2-carboxylic acid, pubchem15865, buttpark 154\50-31, emcs ;6-maleimidocaproic acid nhs, 6-bromo-2-imidazo 1,2-a pyridinecarboxylic acid, 6-bromoimidazo 1,2-a pyridine-2-carboxylicacid
PubChem CID 2757018
IUPAC Name 6-bromoimidazo[1,2-a]pyridine-2-carboxylic acid
SMILES OC(=O)C1=CN2C=C(Br)C=CC2=N1

Specifications

Quantity 250 mg
Formula Weight 241.05 (Anhydrous)
Percent Purity 95%
Chemical Name or Material 6-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'