Learn More
Univet™ 506UP Clear Safety Glasses
The 506 UP features an enveloping panoramic lens for optimum side protection with anti-scratch and anti-fog coating. Temples with rubber padding and adjustment of length and nose bridge ensure wearing comfort.
Brand: Univet™ 506U.03.13.00
This item is not returnable.
View return policy
Description
- Lens available with Vanguard PLUS coating
- Optimal comfort and fit thanks to the soft adjustable nose pad
- Soft terminal tips to eliminate local pressure
Specifications
Rectangular | |
None | |
Polycarbonate | |
Polycarbonate | |
UV380 | |
Not Autoclavable |
Anti-fog, Scratch-resistant | |
Safety Glasses | |
Reusable | |
Polycarbonate | |
Adjustable Temples |