missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B Braun™ 5 L Helipur™ Manual Cleaner and Disinfectant
Click to view available options
Volume (Metric):
5 L
Unit Size:
5 L
Description
- Highly effective liquid disinfection concentrate
- Suitable for surgical instruments made of stainless steel, glass and ceramic
- Cleaning and disinfection in one step; contaminated instruments can be soaked directly in the ready-to-use working solution, manual pre-cleaning can be omitted
- Aldehyde-free
- Economical
- Effective against bacteria (including MRSA and TbB), fungi and enveloped viruses (including HBV, HCV, HIV) and Polyoma- and Adenovirus
- Can be used in ultrasonic baths
Specifications
Specifications
| Product Type | Cleaner |
| Appearance | Red-Brown |
| Container Type | Canister |
| For Use With (Application) | Cleaning and disinfecting of medical devices |
| Volume (Metric) | 5 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction