Learn More
5-Chloro-2'-deoxyuridine, MedChemExpress
MedChemExpress 5-Chloro-2'-deoxyuridine, a thymine analog, is to study the potential of hypochlorous acid damage to DNA and DNA precursors.
Brand: Medchem Express HY-112669
This item is not returnable.
View return policy
Description
5-Chloro-2'-deoxyuridine, a thymine analog, is to study the potential of hypochlorous acid damage to DNA and DNA precursors.Specifications
5-Chloro-2'-deoxyuridine | |
White | |
C9H11ClN2O5 | |
5-Chlorodeoxyuridine CldU | |
OC[C@@H]1[C@H](C[C@H](N2C(NC(C(Cl)=C2)=O)=O)O1)O | |
262.65 | |
Research | |
Neuroscience-Neuromodulation |
50-90-8 | |
Research | |
100 mg | |
DMSO : 250 mg/mL (951.84 mM; Need ultrasonic) | |
262.65 | |
99.18% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only