Learn More
5-Chloro-2'-deoxyuridine, MedChemExpress
MedChemExpress 5-Chloro-2'-deoxyuridine, a thymine analog, is to study the potential of hypochlorous acid damage to DNA and DNA precursors.
Brand:  Medchem Express HY-112669
Encompass
 View more versions of this product
Product Code. 16405516
  • € 136.50 / 100mg
Explore more special offers
Description
Description
5-Chloro-2'-deoxyuridine, a thymine analog, is to study the potential of hypochlorous acid damage to DNA and DNA precursors.
Specifications
Specifications
5-Chloro-2'-deoxyuridine
White
C9H11ClN2O5
5-Chlorodeoxyuridine CldU
OC[C@@H]1[C@H](C[C@H](N2C(NC(C(Cl)=C2)=O)=O)O1)O
262.65
Research
Neuroscience-Neuromodulation
50-90-8
Research
100 mg
DMSO : 250 mg/mL (951.84 mM; Need ultrasonic)
262.65
99.18%
Solid
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only