missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-Chloro-3-Indolyl-β-D-Galactopyranoside, ≥98%, Molecular Biology Reagent, MP Biomedicals™

Product Code. 11452151 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
This item is not returnable. View return policy

Product Code. 11452151

Brand: MP Biomedicals 0219481180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-Bromo-4-Chloro-3-Indolyl-β-D-Galactopyranoside is used as indigogenic substrate for β-galactosidase, for detection of β-galactosidase-positive clones, and the identification of lac and bacterial colonies or phage plaques

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7240-90-6
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.629
MDL Number MFCD00005666
InChI Key OPIFSICVWOWJMJ-AEOCFKNESA-N
Synonym x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
PubChem CID 65181
ChEBI CHEBI:75055
IUPAC Name (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br

Specifications

Color White
Assay Percent Range ≥98%
Quantity 100 mg
Formula Weight 408.6
Grade Molecular Biology Reagent
Physical Form Powder
Chemical Name or Material 5-Bromo-4-Chloro-3-Indoyl-β-D-Galactopyranoside, FTIR
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.