Learn More
DWK Life Sciences Kimble Chase™ 5.1 Borosilicate Glass Disposable Test Tubes
Click to view available options
Length (Metric):
70 mm
75 mm
100 mm
130 mm
180 mm
200 mm
Diameter (Metric):
8 mm
10 mm
12 mm
14 mm
16 mm
18 mm
20 mm
25 mm
30 mm
Unit Size:
Pack of 100
Pack of 50
Description
- Disposable
- Clear borosilicate glass
- Round bottom
- Rimmed
Specifications
Specifications
| Design Type | Round bottom |
| Length (Metric) | 75 mm |
| Packaging | 100Pack |
| Rimmed | Yes |
| Diameter (Metric) | 12 mm |
| Product Type | Test Tube |