Learn More
Brady™ IP Printer Enabled 4400 Series Thermal Transfer Printer Ribbons
High density ribbon offers solvent, high heat and scratch resistance
Brand: Brady™ 066047
This item is not returnable.
View return policy
Description
- Includes spindle
- Compatible with IP300, IP600 printers
- Core inside diameter: 25.40mm (1 in.), ink side out
Specifications
Thermal Transfer Printer Ribbon | |
IP300 Thermal Transfer Printer, IP600 Thermal Transfer Printer | |
984 in. | |
300 m | |
25.4 mm I.D. | |
Blue 4400 Series Thermal Transfer Printer Ribbon - Brady IP Printer Enabled | |
Ink side out |
Blue | |
Resin | |
2.36 in. | |
5.99 cm | |
300 m x 5.99 cm (984 ft. x 2.36 in.) | |
IP™ Enabled | |
1 Ea. |