Learn More
Schneider Gerd™ 4301 Stainless Steel Double Ended Spoon
Product Code. p-8008612
Click to view available options
Length (Metric):
120 mm
180 mm
250 mm
300 mm
Unit Size:
Each
Product Code. 10672902
Brand: Schneider Gerd™ 231
Only null left
Add to basket
Add to basket
Use this spoon for a variety of applications. Schneider Gerd™ 4301 Stainless Steel Double Ended Spoons are ideal for use with a wide range of compounds.
  • Spoons collect and transport dry substances or liquids
  • Two end shapes offer increased versatility
  • Ideal for working with powders, stirring and mixing
  • Perfect for miniscule amounts of chemicals
Specifications
Quantity 1 Pk.
Product Type Double Spoon End Spatula
For Use With (Application) For Chemical
Material Stainless Steel (4301)
Length (Metric) 120 mm