missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl bromoacetate, 98+%, Thermo Scientific™

Product Code. 11317975
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11317975

Brand: Thermo Scientific Alfa Aesar L08058.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16869-24-2
Molecular Formula C9H8BrNO4
Molecular Weight (g/mol) 274.07
MDL Number MFCD00051838
InChI Key ADHFTAKIDKDGBV-UHFFFAOYSA-N
Synonym 4-nitrobenzyl bromoacetate, 4-nitrobenzyl 2-bromoacetate, p-nitrobenzyl bromoacetate, bromoacetic acid-4-nitrobenzylester, acetic acid, bromo-, 4-nitrophenyl methyl ester, 4-nitrophenyl methyl 2-bromoacetate, acmc-1c4y8, p-nitrobenzyl alpha-bromoacetate, bromoacetic acid 4-nitrobenzyl ester
PubChem CID 7015321
IUPAC Name (4-nitrophenyl)methyl 2-bromoacetate
SMILES C1=CC(=CC=C1COC(=O)CBr)[N+](=O)[O-]

Specifications

Melting Point 48°C to 52°C
Boiling Point 105°C to 106°C (5 mmHg)
Quantity 50 g
Beilstein 2531330
Formula Weight 274.08
Percent Purity ≥98%
Chemical Name or Material 4-Nitrobenzyl bromoacetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.