missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzylsulfonyl chloride, 97%

Product Code. p-7077513 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15203558

Brand: Thermo Scientific Acros 454840010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 51419-59-1 | C8H9ClO2S | 204.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51419-59-1
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.68
InChI Key JALKUHLLMWYIAT-UHFFFAOYSA-N
Synonym 4-methylbenzylsulfonyl chloride, 4-methylphenyl methanesulfonyl chloride, p-tolylmethanesulfonyl chloride, p-tolyl-methanesulfonyl chloride, p-tolyl methanesulfonyl chloride, 4-methylphenyl methyl sulfonyl chloride, pubchem5592, pubchem5631, acmc-209wgt, 4-methylbenzylsulfonylchloride
PubChem CID 2794648
IUPAC Name (4-methylphenyl)methanesulfonyl chloride
SMILES CC1=CC=C(C=C1)CS(=O)(=O)Cl

Specifications

Melting Point 78.0°C to 82.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 204.68
Percent Purity 97%
Chemical Name or Material 4-Methylbenzylsulfonyl chloride
compliance-icons compliance-icons
Product Identifier
  • 4-Methylbenzylsulfonyl chloride, 97%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.