Learn More
4'-Hydroxynonanophenone, 96%
Product Code. 11347655
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11347655
Brand: Thermo Scientific Alfa Aesar L02293.06
Request Bulk or Custom Format
CAS: 14392-69-9 | C15H22O2 | 234.34 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14392-69-9
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.34
MDL Number MFCD00020174
InChI Key JZBXYOOARNRUME-UHFFFAOYSA-N
Synonym 1-4-hydroxyphenyl-1-nonanone, 1-4-hydroxyphenyl nonan-1-one, p-hydroxynonanophenone, 4'-hydroxynonanophenone, pelargonyl-4-phenol, 1-nonanone,1-4-hydroxyphenyl, 1-nonanone, 1-4-hydroxyphenyl
PubChem CID 167093
IUPAC Name 1-(4-hydroxyphenyl)nonan-1-one
SMILES CCCCCCCCC(=O)C1=CC=C(O)C=C1
Specifications
Melting Point 53°C to 57°C
Quantity 5 g
Beilstein 2103401
Formula Weight 234.34
Percent Purity 96%
Chemical Name or Material 4'-Hydroxynonanophenone