missing translation for 'onlineSavingsMsg'
Learn More
4'-Fluoro-3'-methoxyacetophenone, 98%, Thermo Scientific Chemicals Product Code.: 11309925

4'-Fluoro-3'-methoxyacetophenone, 98%, Thermo Scientific Chemicals

Product Code. 11309925
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11309925

Brand: Thermo Scientific Alfa Aesar L19894.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64287-19-0
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD00272135
InChI Key PFEGFUCYOHBDJF-UHFFFAOYSA-N
Synonym 1-4-fluoro-3-methoxyphenyl ethanone, 4'-fluoro-3'-methoxyacetophenone, 4-fluoro-3-methoxyacetophenone, ethanone, 1-4-fluoro-3-methoxyphenyl, 1-4-fluoro-3-methoxyphenyl ethan-1-one, 1-4-fluoro-3-methoxy-phenyl-ethanone, 1-acetyl-4-fluoro-3-methoxybenzene, pubchem4234, ksc621a4h, attercop-chm at126609Show MoreShow Less
PubChem CID 2774532
IUPAC Name 1-(4-fluoro-3-methoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)F)OC

Specifications

Melting Point 51°C to 53°C
Quantity 250 mg
Beilstein 4381319
Formula Weight 168.17
Percent Purity 98%
Chemical Name or Material 4'-Fluoro-3'-methoxyacetophenone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10125104 Certificate TypeCertificate of Analysis Date01/05/2023 Catalog NumberL19894.03,L19894.MD
Lot Number10125104 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL19894.03,L19894.MD
Lot Number10125104 Certificate TypeCertificate of Analysis Date12/08/2020 Catalog NumberL19894.03,L19894.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4'-Fluoro-3'-methoxyacetophenone, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.