missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxybenzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 11386959
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Carboxybenzeneboronic acid pinacol ester is used to improvement of transition metal-mediated carbon-carbon as well as carbon-heteroatom cross-coupling reactions as a labeling feature with the focus on the preparation of radio labeled compounds for molecular imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Carboxybenzeneboronic acid pinacol ester is used to improvement of transition metal-mediated carbon-carbon as well as carbon-heteroatom cross-coupling reactions as a labeling feature with the focus on the preparation of radio labeled compounds for molecular imaging.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180516-87-4
Molecular Formula C13H17BO4
Molecular Weight (g/mol) 248.09
MDL Number MFCD01863710
InChI Key IYDKBQIEOBXLTP-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 4-carboxyphenylboronic acid pinacol ester, 4-carboxylphenylboronic acid pinacol ester, 4-carboxyphenylboronic acid, pinacol ester, 4-carboxyphenylboronicacidpinacolester, 4-carboxybenzeneboronic acid pinacol ester, 4-carboxyphenylboronic acid pinacolate, 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 4-carboxyphenylboronic acid,pinacol ester, 4-carboxyphenyl boronic acid, pinacol ester
PubChem CID 2734621
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 227°C to 231°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 248.08
Percent Purity 97%
Chemical Name or Material 4-Carboxybenzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'